Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dibromo-2,3-dichloroaniline, 98%

$24.55 - $92.66

Chemical Identifiers

Chemical Name or Material 4,6-Dibromo-2,3-dichloroaniline
CAS 113571-15-6
Molecular Formula C6H3Br2Cl2N
Formula Weight 319.82
MDL Number MFCD00052815
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1496103

  SDS
alfa aesar™
A1496103
1g Each for $24.55

AAA1496106

  SDS
alfa aesar™
A1496106
5g Each for $92.66
Description & Specifications

Specifications

Formula Weight 319.82
Chemical Name or Material 4,6-Dibromo-2,3-dichloroaniline
Melting Point 96° to 98°C
Assay 98%
Quantity 1g
Molecular Formula C6H3Br2Cl2N
MDL Number MFCD00052815
CAS 113571-15-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.