missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dibromo-2,3-dichloroaniline, 98%

Manufacturer:  Alfa Aesar™ A1496103

 View more versions of this product

Catalog No. AAA1496103



Specifications

Specifications

4,6-Dibromo-2,3-dichloroaniline
1g
C6H3Br2Cl2N
benzenamine,4,6-dibromo-2,3-dichloro, acmc-20amrl, 4,6-dibromo-2,3-dichlorophenylamine, 4,6-dibromo-2,3-dichloro-phenylamine, 4,6-dibromo-2,3-dichloroaniline, 4,6-bis bromanyl-2,3-bis chloranyl aniline
C1=C(C(=C(C(=C1Br)Cl)Cl)N)Br
319.805
319.82
96°C to 98°C
113571-15-6
MFCD00052815
IPWGDMQXLGFBLC-UHFFFAOYSA-N
4,6-dibromo-2,3-dichloroaniline
2776953
98%

Chemical Identifiers

113571-15-6
319.805
IPWGDMQXLGFBLC-UHFFFAOYSA-N
2776953
C1=C(C(=C(C(=C1Br)Cl)Cl)N)Br
C6H3Br2Cl2N
MFCD00052815
benzenamine,4,6-dibromo-2,3-dichloro, acmc-20amrl, 4,6-dibromo-2,3-dichlorophenylamine, 4,6-dibromo-2,3-dichloro-phenylamine, 4,6-dibromo-2,3-dichloroaniline, 4,6-bis bromanyl-2,3-bis chloranyl aniline
4,6-dibromo-2,3-dichloroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents