Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6'-Dibromo-2,3'-bipyridine, 95%

$174.78 - $622.30

Chemical Identifiers

Chemical Name or Material 4,6'-Dibromo-2,3'-bipyridine
CAS 942206-14-6
Molecular Formula C10H6Br2N2
Formula Weight 313.98
MDL Number MFCD09037425
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27679MC

  SDS
alfa aesar™
H27679MC
100mg Each for $174.78

AAH27679ME

  SDS
alfa aesar™
H27679ME
500mg Each for $622.30
Description & Specifications

Specifications

Formula Weight 313.98
Melting Point 127° to 130°C
Molecular Formula C10H6Br2N2
CAS 942206-14-6
Health Hazard 1 Warning
Assay 95%
Quantity 100mg
MDL Number MFCD09037425
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4,6'-Dibromo-2,3'-bipyridine
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.