missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Diaminoresorcinol dihydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC333671000

 View more versions of this product

Catalog No. AC333671000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4, 6-Diaminoresorcinol dihydrochloride
100g
97.5
Authentic
Crystalline Solid
98%
213.06
13, II, 474
Gray-brown
16523-31-2
100.0
Glass bottle
≥97.5%
C6H8N2O2·2HCl
MFCD00143239

Chemical Identifiers

16523-31-2
100.0
C6H8N2O2·2HCl
MFCD00143239
≥97.5%
97.5
98%
213.06
4, 6-Diaminoresorcinol dihydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.