Please login to your online account to display your discounted pricing

4,6-Diaminoresorcinol dihydrochloride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC333670250

 View more versions of this product

Catalog No. AC333670250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Solid
Formula Weight 213.06
Quantity 25g
Color Gray-brown
Beilstein 13, II, 474
CAS Max % 100.0
Chemical Name or Material 4, 6-Diaminoresorcinol dihydrochloride
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H8N2O2·2HCl
MDL Number MFCD00143239
CAS 16523-31-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4, 6-Diaminoresorcinol dihydrochloride
CAS 16523-31-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H8N2O2·2HCl
Formula Weight 213.06
MDL Number MFCD00143239
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents