missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Diaminoresorcinol dihydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 333670050

 View more versions of this product

Catalog No. AC333670050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
213.06
Gray-brown
100.0
MFCD00143239
16523-31-2
Glass bottle
C6H8N2O2·2HCl
Crystalline Solid
5g
13, II, 474
Authentic
98%
97.5
4, 6-Diaminoresorcinol dihydrochloride

Chemical Identifiers

16523-31-2
100.0
C6H8N2O2·2HCl
MFCD00143239
≥97.5%
97.5
98%
213.06
4, 6-Diaminoresorcinol dihydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.