missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,6-Diaminopyrimidine Hemisulfate, Monohydrate 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 112410050
Specifications
4,6-Diaminopyrimidine hemisulfate monohydrate | |
Glass Bottle | |
243.0°C to 244.0°C | |
98% | |
97.5 | |
98% | |
MFCD00149655 | |
ZMRLIHKEUKNTRB-UHFFFAOYSA-N | |
pyrimidine-4,6-diamine;sulfuric acid;hydrate | |
54772454 | |
Crystalline Powder |
Authentic | |
Yellow | |
5g | |
207226-28-6,77709-02-5 | |
100.0 | |
C8H16N8O5S | |
4,6-diaminopyrimidine hemisulfate salt monohydrate, bis pyrimidine-4,6-diamine sulfuric acid hydrate, sulfuric acid-pyrimidine-4,6-diamine-water 1/2/1 | |
C1=C(N=CN=C1N)N.C1=C(N=CN=C1N)N.O.OS(=O)(=O)O | |
336.327 | |
177.18 | |
98% |
Chemical Identifiers
207226-28-6 | |
336.327 | |
ZMRLIHKEUKNTRB-UHFFFAOYSA-N | |
54772454 | |
C1=C(N=CN=C1N)N.C1=C(N=CN=C1N)N.O.OS(=O)(=O)O |
C8H16N8O5S | |
MFCD00149655 | |
4,6-diaminopyrimidine hemisulfate salt monohydrate, bis pyrimidine-4,6-diamine sulfuric acid hydrate, sulfuric acid-pyrimidine-4,6-diamine-water 1/2/1 | |
pyrimidine-4,6-diamine;sulfuric acid;hydrate |
Safety and Handling
EINECSNumber : 278-754-7