Please login to your online account to display your discounted pricing

4,6-Diaminopyrimidine Hemisulfate, Monohydrate 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  112410010

 View more versions of this product

Catalog No. AC112410010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 177.18
Quantity 1g
Color Yellow
EINECS Number 278-754-7
CAS Max % 100.0
Chemical Name or Material 4, 6-Diaminopyrimidine hemisulfate monohydrate
Packaging Glass bottle
Name Note 98%
Molecular Formula 5 H2SO4·H2O
MDL Number MFCD00149655
CAS 207226-28-6
CAS Min % 97.5
CAS 77709-02-5
Infrared Spectrum Authentic
Assay Percent Range 98%

Specifications

Chemical Name or Material 4, 6-Diaminopyrimidine hemisulfate monohydrate
Name Note 98%
CAS 207226-28-6
CAS Min % 97.5
CAS Max % 100.0
CAS 77709-02-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula 5 H2SO4·H2O
Formula Weight 177.18
MDL Number MFCD00149655