missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Diaminopyrimidine Hemisulfate, Monohydrate 98%, ACROS Organics™

$85.94 - $85.94

Chemical Identifiers

CAS 207226-28-6,77709-02-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula 5 H2SO4·H2O
Formula Weight 177.18
MDL Number MFCD00149655
Chemical Name or Material 4, 6-Diaminopyrimidine hemisulfate monohydrate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC112410050 SDS Fisher Scientific Acros
112410050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC112410010 SDS Thermo Fisher Scientific
112410010
Glass bottle 1g Each for $85.94
Specifications Description & Specifications

Chemical Identifiers

207226-28-6,77709-02-5
100.0
5 H2SO4·H2O
MFCD00149655
≥97.5%
97.5
98%
177.18
4, 6-Diaminopyrimidine hemisulfate monohydrate, 98%

Specifications

177.18
5g
100.0
97.5
Authentic
5 H2SO4·H2O
4, 6-Diaminopyrimidine hemisulfate monohydrate, 98%
Crystalline Powder
Yellow
Glass bottle
77709-02-5,207226-28-6
MFCD00149655
98%
≥97.5%

Safety and Handling

EINECSNumber : 278-754-7