Learn More
4,6-Diamino-2-(methylthio)pyrimidine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0788814
Description
4,6-Diamino-2-(methylthio)pyrimidine can reacts with 3-Ethoxy-acrylic acid ethyl ester to get 4-Amino-2-methylsulfanyl-8H-pyrido[2,3-d]pyrimidin-7-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,6-Diamino-2-(methylthio)pyrimidine can reacts with 3-Ethoxy-acrylic acid ethyl ester to get 4-Amino-2-methylsulfanyl-8H-pyrido[2,3-d]pyrimidin-7-one.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
4,6-Diamino-2-(methylthio)pyrimidine | |
187°C to 191°C | |
MFCD00023241 | |
127235 | |
AHAIUNAIAHSWPG-UHFFFAOYSA-N | |
2-methylsulfanylpyrimidine-4,6-diamine | |
70497 | |
97% |
1005-39-6 | |
C5H8N4S | |
25 g | |
4,6-diamino-2-methylmercaptopyrimidine, 2-methylthio pyrimidine-4,6-diamine, 2-methylthiopyrimidine-4,6-diamine, 4,6-diamino-2-methylthiopyrimidine, 4,6-pyrimidinediamine, 2-methylthio, 4,6-diamino-2-methylthio pyrimidine, 2-methylsulfanyl pyrimidine-4,6-diamine, 2-methylthio-4,6-diaminopyrimidine, 4,6-diamino-2-methyl-mercaptopyrimidine, pyrimidine, 4,6-diamino-2-methylthio | |
CSC1=NC(=CC(=N1)N)N | |
156.207 | |
156.21 |
Chemical Identifiers
1005-39-6 | |
156.207 | |
AHAIUNAIAHSWPG-UHFFFAOYSA-N | |
70497 | |
CSC1=NC(=CC(=N1)N)N |
C5H8N4S | |
MFCD00023241 | |
4,6-diamino-2-methylmercaptopyrimidine, 2-methylthio pyrimidine-4,6-diamine, 2-methylthiopyrimidine-4,6-diamine, 4,6-diamino-2-methylthiopyrimidine, 4,6-pyrimidinediamine, 2-methylthio, 4,6-diamino-2-methylthio pyrimidine, 2-methylsulfanyl pyrimidine-4,6-diamine, 2-methylthio-4,6-diaminopyrimidine, 4,6-diamino-2-methyl-mercaptopyrimidine, pyrimidine, 4,6-diamino-2-methylthio | |
2-methylsulfanylpyrimidine-4,6-diamine |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 213-735-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only