missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202710100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4′,6′-Diamidino-2-phenylindole dihydrochloride | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00012681 | |
4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836 | |
FPNZBYLXNYPRLR-UHFFFAOYSA-N | |
2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide;dihydrochloride | |
160166 | |
98% |
28718-90-3 | |
100.0 | |
97.5% min. (HPLC) | |
C16H17Cl2N5 | |
10 mg | |
(1% in water) Clear | |
[H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N | |
350.25 | |
350.25 | |
Solution |
Chemical Identifiers
28718-90-3 | |
350.25 | |
FPNZBYLXNYPRLR-UHFFFAOYSA-N | |
160166 |
C16H17Cl2N5 | |
MFCD00012681 | |
4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836 | |
[H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N |
Safety and Handling
EINECSNumber : 249-186-7
RUO – Research Use Only