Learn More
4,6-Decadiyne, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5346706
Description
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.
Solubility
Immiscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
4,6-Decadiyne | |
0.82 | |
C10H14 | |
MFCD00041645 | |
4,6-decadiyne, acmc-1c48i | |
LIWZSNTUMSGWTF-UHFFFAOYSA-N | |
deca-4,6-diyne | |
140067 | |
97% |
16387-71-6 | |
96°C (15 mmHg) | |
CH3(CH2)2C≡CC≡C(CH2)2CH3 | |
5 g | |
Immiscible with water. | |
CCCC#CC#CCCC | |
134.222 | |
134.22 |
Chemical Identifiers
16387-71-6 | |
134.222 | |
LIWZSNTUMSGWTF-UHFFFAOYSA-N | |
140067 | |
CCCC#CC#CCCC |
C10H14 | |
MFCD00041645 | |
4,6-decadiyne, acmc-1c48i | |
deca-4,6-diyne |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only