missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(6-Bromopyrid-2-yl)morpholine, ≥97%, Maybridge

Chemical Identifiers

CAS 332134-60-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H11BrN2O
Formula Weight 243.1
MDL Number MFCD07772824
Chemical Name or Material 4-(6-Bromopyrid-2-yl)morpholine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC38310DA SDS Fisher Scientific Company
CC38310DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

332134-60-8
100.0
C9H11BrN2O
MFCD07772824
97%
96.5
97%
243.1
4-(6-Bromopyrid-2-yl)morpholine

Specifications

332134-60-8
100.0
1g
C9H11BrN2O
MFCD07772824
97%
96.5
Amber Glass Bottle
97%
243.1
4-(6-Bromopyrid-2-yl)morpholine

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning