missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Bis(diphenylphosphino)phenoxazine, TCI America™

$68.75 - $331.53

Chemical Identifiers

CAS 261733-18-0
Molecular Formula C36H27NOP2
Molecular Weight (g/mol) 551.566
MDL Number MFCD03788937
InChI Key HSWZLYXRAOXOLL-UHFFFAOYSA-N
Synonym 4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
PubChem CID 3255346
IUPAC Name (6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC4=C3OC5=C(N4)C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B2717100MG View Documents TCI America
B2717100MG
100mg Each for $68.75
 
B27171G View Documents TCI America
B27171G
1g Each for $331.53
 
Specifications

Chemical Identifiers

261733-18-0
551.566
HSWZLYXRAOXOLL-UHFFFAOYSA-N
3255346
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC4=C3OC5=C(N4)C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
C36H27NOP2
MFCD03788937
4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
(6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane

Specifications

White-Yellow
261733-18-0
MFCD03788937
HSWZLYXRAOXOLL-UHFFFAOYSA-N
(6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane
3255346
Crystal-Powder at 20°C
4,6-Bis(diphenylphosphino)phenoxazine
100mg
C36H27NOP2
4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC4=C3OC5=C(N4)C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
551.566
551.57
≥98.0% (HPLC,N)
Safety and Handling

Safety and Handling

TSCA : No