Learn More
4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5431503
Description
4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
4-(6-Amino-3-pyridyl)-1-Boc-piperazine | |
C14H22N4O2 | |
1 g | |
Air sensitive | |
RMULRXHUNOVPEI-UHFFFAOYSA-N | |
tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate | |
11737525 | |
97% |
571188-59-5 | |
MFCD11594962 | |
10173494 | |
tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester | |
CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 | |
278.36 | |
278.35 |
Chemical Identifiers
571188-59-5 | |
278.36 | |
RMULRXHUNOVPEI-UHFFFAOYSA-N | |
tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate |
C14H22N4O2 | |
MFCD11594962 | |
11737525 | |
CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only