missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%

Manufacturer:  Alfa Aesar™ H54315MD

 View more versions of this product

Catalog No. AAH54315MD



Description

Description

Applications
4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-(6-Amino-3-pyridyl)-1-Boc-piperazine
250mg
C14H22N4O2
10173494
RMULRXHUNOVPEI-UHFFFAOYSA-N
tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
11737525
97%
Air sensitive
571188-59-5
MFCD11594962
1-boc-4-6-amino-3-pyridyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CN=C(C=C2)N
278.356
278.35

Chemical Identifiers

571188-59-5
278.356
RMULRXHUNOVPEI-UHFFFAOYSA-N
11737525
CC(C)(C)OC(=O)N1CCN(CC1)C2=CN=C(C=C2)N
C14H22N4O2
MFCD11594962
1-boc-4-6-amino-3-pyridyl piperazine
tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents