missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[5-(Trifluoromethyl)pyridin-2-yl]benzaldehyde, 90%, Thermo Scientific™

Chemical Identifiers

CAS 871252-64-1
Molecular Formula C13H8F3NO
Molecular Weight (g/mol) 251.208
MDL Number MFCD06797372
InChI Key SBXWMOPSHGIWEN-UHFFFAOYSA-N
Synonym 4-5-trifluoromethyl pyridin-2-yl benzaldehyde, 4-5-trifluoromethyl-2-pyridinyl benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridinyl-benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridyl benzaldehyde, 4-5-trifluoromethyl-pyridin-2-yl-benzaldehyde, benzaldehyde,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl pyridin-2-yl benzenecarbaldehyde
PubChem CID 2763359
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=NC=C(C=C2)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC53004CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC53004CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC53004DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC53004DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

871252-64-1
251.208
SBXWMOPSHGIWEN-UHFFFAOYSA-N
2763359
C1=CC(=CC=C1C=O)C2=NC=C(C=C2)C(F)(F)F
C13H8F3NO
MFCD06797372
4-5-trifluoromethyl pyridin-2-yl benzaldehyde, 4-5-trifluoromethyl-2-pyridinyl benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridinyl-benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridyl benzaldehyde, 4-5-trifluoromethyl-pyridin-2-yl-benzaldehyde, benzaldehyde,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl pyridin-2-yl benzenecarbaldehyde
4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde

Specifications

871252-64-1
100.0
Amber Glass Bottle
MFCD06797372
4-5-trifluoromethyl pyridin-2-yl benzaldehyde, 4-5-trifluoromethyl-2-pyridinyl benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridinyl-benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridyl benzaldehyde, 4-5-trifluoromethyl-pyridin-2-yl-benzaldehyde, benzaldehyde,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl pyridin-2-yl benzenecarbaldehyde
C1=CC(=CC=C1C=O)C2=NC=C(C=C2)C(F)(F)F
251.208
251.21
4-[5-(Trifluoromethyl)pyridin-2-yl]benzaldehyde
89.5
90%
C13H8F3NO
250mg
SBXWMOPSHGIWEN-UHFFFAOYSA-N
4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
2763359
90%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'