Please login to your online account to display your discounted pricing

{4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC53013CB

 View more versions of this product

Catalog No. CC53013CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 252.24
Quantity 250mg
Assay 97%
CAS Max % 100.0
Molecular Formula C13H11F3N2
Chemical Name or Material {4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine
MDL Number MFCD09817478
CAS 906352-74-7
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.

Specifications

Chemical Name or Material {4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine
CAS 906352-74-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H11F3N2
Formula Weight 252.24
MDL Number MFCD09817478
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents