Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Methylenedioxy-2-nitrocinnamic acid, 98%

$147.31 - $554.14

Chemical Identifiers

Chemical Name or Material 4,5-Methylenedioxy-2-nitrocinnamic acid
CAS 6315-90-8
Molecular Formula C10H7NO6
Formula Weight 237.17
MDL Number MFCD00063250
Beilstein 88864
Synonym 3-(6-Nitro-1,3-benzodioxol-5-yl)propenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0801406

 
alfa aesar™
L0801406
5g Each for $147.31

AAL0801414

 
alfa aesar™
L0801414
25g Each for $554.14
Description & Specifications

Specifications

Formula Weight 237.17
Chemical Name or Material 4,5-Methylenedioxy-2-nitrocinnamic acid
Molecular Formula C10H7NO6
MDL Number MFCD00063250
CAS 6315-90-8
Beilstein 88864
Synonym 3-(6-Nitro-1,3-benzodioxol-5-yl)propenoic acid
Health Hazard 1 Warning
Assay 98%
Melting Point ≈280°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.