missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4',5'-Methylenedioxy-2'-nitroacetophenone, 98%

Manufacturer:  Alfa Aesar™ L1184106

 View more versions of this product

Catalog No. AAL1184106



Description

Description

Applications
4′,5′-Methylenedioxy-2′-nitroacetophenone is used as an intermediate in organic syntheses and in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4',5'-Methylenedioxy-2'-nitroacetophenone
5g
C9H7NO5
199756
BQONDGIXVHVIIR-UHFFFAOYSA-N
1-(6-nitro-1,3-benzodioxol-5-yl)ethanone
92022
98%
121°C to 123°C
56136-84-6
MFCD00053045
1-6-nitro-1,3-benzodioxol-5-yl ethanone, 4',5'-methylenedioxy-2'-nitroacetophenone, 4,5-methylenedioxy-2-nitroacetophenone, 1-6-nitrobenzo d 1,3 dioxol-5-yl ethanone, 1-6-nitro-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethanone, 5-acetyl-6-nitro-1,3-benzodioxole, 4.5-methylenedioxy-2-nitroacetophenone, 5-acetyl-6-nitro-2h-benzo d 1,3-dioxolene
CC(=O)C1=CC2=C(C=C1[N+](=O)[O-])OCO2
209.157
209.16

Chemical Identifiers

56136-84-6
209.157
BQONDGIXVHVIIR-UHFFFAOYSA-N
92022
CC(=O)C1=CC2=C(C=C1[N+](=O)[O-])OCO2
C9H7NO5
MFCD00053045
1-6-nitro-1,3-benzodioxol-5-yl ethanone, 4',5'-methylenedioxy-2'-nitroacetophenone, 4,5-methylenedioxy-2-nitroacetophenone, 1-6-nitrobenzo d 1,3 dioxol-5-yl ethanone, 1-6-nitro-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethanone, 5-acetyl-6-nitro-1,3-benzodioxole, 4.5-methylenedioxy-2-nitroacetophenone, 5-acetyl-6-nitro-2h-benzo d 1,3-dioxolene
1-(6-nitro-1,3-benzodioxol-5-yl)ethanone
Documents