Please login to your online account to display your discounted pricing

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride, 97%, Maybridge

$475.86 - $475.86

Chemical Identifiers

Chemical Name or Material 4-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzoyl chloride
CAS 222541-76-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7ClN2O2
Formula Weight 222.63
MDL Number MFCD07368530
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC34802DA

 
fisher scientific company
CC34802DA
Amber Glass Bottle 1g Each for $475.86
Description & Specifications

Specifications

Quantity 1g
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Chemical Name or Material 4-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzoyl chloride
Packaging Amber Glass Bottle
Assay 97%
Molecular Formula C10H7ClN2O2
Formula Weight 222.63
MDL Number MFCD07368530
CAS 222541-76-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
CAS Min % 96.5
CAS Max % 100.0
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.