missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC34802DA

 View more versions of this product

Catalog No. CC34802DA


 

Specifications Description & Specifications

Specifications

1g
222.63
C10H7ClN2O2
MFCD07368530
97%
100.0
4-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzoyl chloride
97%
222541-76-6
Amber Glass Bottle
96.5

Chemical Identifiers

222541-76-6
100.0
C10H7ClN2O2
MFCD07368530
97%
96.5
97%
222.63
4-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzoyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger