Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(5-Iodo-2-pyridyl)morpholine, 96%

$258.59 - $258.59

Chemical Identifiers

Chemical Name or Material 4-(5-Iodo-2-pyridyl)morpholine
CAS 470463-42-4
Molecular Formula C9H11IN2O
Formula Weight 290.11
MDL Number MFCD04039866
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5014203

 
alfa aesar™
H5014203
1g Each for $258.59
Description & Specifications

Specifications

Formula Weight 290.11
Chemical Name or Material 4-(5-Iodo-2-pyridyl)morpholine
Assay 96%
Molecular Formula C9H11IN2O
MDL Number MFCD04039866
CAS 470463-42-4
Melting Point 131° to 135°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.