Please login to your online account to display your discounted pricing

Alfa Aesar™ 4(5)-Imidazolemethanol hydrochloride, 98+%

$42.50 - $364.64

Chemical Identifiers

Chemical Name or Material 4(5)-Imidazolemethanol hydrochloride
CAS 32673-41-9
Molecular Formula C4H6N2O·HCl
Formula Weight 134.57
MDL Number MFCD00012697
Beilstein 3562041
Synonym 4(5)-(Hydroxymethyl)imidazole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2018203

  SDS
alfa aesar™
L2018203
1g Each for $42.50

AAL2018206

  SDS
alfa aesar™
L2018206
5g Each for $142.39

AAL2018214

  SDS
alfa aesar™
L2018214
25g Each for $364.64
Description & Specifications

Specifications

Formula Weight 134.57
Chemical Name or Material 4(5)-Imidazolemethanol hydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 4(5)-(Hydroxymethyl)imidazole hydrochloride
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 32673-41-9
Health Hazard 1 Warning
Molecular Formula C4H6N2O·HCl
Assay ≥98%
MDL Number MFCD00012697
Recommended Storage 2° to 8°C
Melting Point 104° to 108°C
Quantity 1g
Beilstein 3562041

Applications
It has been used in the studies on some heterocyclic ligands showing linked ionization effects. 4-(Hydroxymethyl)imidazole HCl has been used in the synthesis of imidazole-containing peptidomimetic inhibitors.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C