missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4(5)-Ethylimidazole, TCI America™

Manufacturer:  TCI America E09841G

 View more versions of this product

Catalog No. E09841G



Specifications

Specifications

4(5)-Ethylimidazole
Yellow
19141-85-6
NJQHZENQKNIRSY-UHFFFAOYSA-N
5-ethyl-1H-imidazole
10153736
Clear Liquid at 20°C
140°C
1g
C5H8N2
CCC1=CN=CN1
96.133
96.13
≥98.0% (GC,T)

Chemical Identifiers

19141-85-6
96.133
10153736
CCC1=CN=CN1
C5H8N2
NJQHZENQKNIRSY-UHFFFAOYSA-N
5-ethyl-1H-imidazole
Safety and Handling

Safety and Handling

TSCA : No

Documents