Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Diphenylimidazolin-2-one, 98%

$52.49 - $169.32

Chemical Identifiers

Chemical Name or Material 4,5-Diphenylimidazolin-2-one
CAS 642-36-4
Molecular Formula C15H12N2O
Formula Weight 236.28
MDL Number MFCD00022393
Beilstein 187611
Synonym 4,5-Diphenyl-2-glyoxalone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0734003

  SDS
alfa aesar™
L0734003
1g Each for $52.49

AAL0734006

  SDS
alfa aesar™
L0734006
5g Each for $169.32
Description & Specifications

Specifications

Formula Weight 236.28
Chemical Name or Material 4,5-Diphenylimidazolin-2-one
Assay 98%
Molecular Formula C15H12N2O
CAS 642-36-4
Beilstein 187611
Synonym 4,5-Diphenyl-2-glyoxalone
Quantity 1g
MDL Number MFCD00022393
Melting Point 323° to 326°C