missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Diphenylimidazole, 98+%

Manufacturer:  Alfa Aesar™ A1493914

 View more versions of this product

Catalog No. AAA1493914



Description

Description

Applications
4,5-Diphenylimidazole is used as a precursor for azo dyes such as 2-(quinolylazo)-4,5-diphenylimidazole . It is also used to determine metal ions by spectrophotometrically. It acts as a chromogenic reagent.

Solubility
Soluble in methanol.

Notes
Incompatible with strong acids.
Specifications

Specifications

4,5-Diphenylimidazole
25g
C15H12N2
157587
CPHGOBGXZQKCKI-UHFFFAOYSA-N
4,5-diphenyl-1H-imidazole
69588
≥98%
230°C to 234°C
668-94-0
MFCD00005198
4,5-diphenylimidazole, diphenylimidazole, 1h-imidazole, 4,5-diphenyl, imidazole, 4,5-diphenyl, dsstox_cid_25700, dsstox_rid_81072, dsstox_gsid_45700, 4,5-diphenyl-1h-imidazol, imidazole,5-diphenyl, 4,5-diphenylglyoxaline
C1=CC=C(C=C1)C2=C(N=CN2)C3=CC=CC=C3
220.275
220.28

Chemical Identifiers

668-94-0
220.275
CPHGOBGXZQKCKI-UHFFFAOYSA-N
69588
C1=CC=C(C=C1)C2=C(N=CN2)C3=CC=CC=C3
C15H12N2
MFCD00005198
4,5-diphenylimidazole, diphenylimidazole, 1h-imidazole, 4,5-diphenyl, imidazole, 4,5-diphenyl, dsstox_cid_25700, dsstox_rid_81072, dsstox_gsid_45700, 4,5-diphenyl-1h-imidazol, imidazole,5-diphenyl, 4,5-diphenylglyoxaline
4,5-diphenyl-1H-imidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents