missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Diphenyl-2-imidazolethiol, 97%, ACROS Organics™

Chemical Identifiers

CAS 2349-58-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C15H12N2S
Formula Weight 252.33
MDL Number MFCD00005189
Chemical Name or Material 4, 5-Diphenyl-2-imidazolethiol, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC155960250 SDS Thermo Fisher Scientific
155960250
Plastic bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2349-58-8
100.0
C15H12N2S
MFCD00005189
≥96.0%
96.0
97%
252.33
4, 5-Diphenyl-2-imidazolethiol, 97%

Specifications

252.33
25g
24, 214
96.0
Authentic
MFCD00005189
C15H12N2S
≥96.0%
Fine Crystalline Powder
White to light yellow
100.0
Plastic bottle
97%
2349-58-8
4, 5-Diphenyl-2-imidazolethiol, 97%