Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Diphenyl-1,2,3-thiadiazole, 97%

$71.43 - $270.92

Chemical Identifiers

Chemical Name or Material 4,5-Diphenyl-1,2,3-thiadiazole
CAS 5393-99-7
Molecular Formula C14H10N2S
Formula Weight 238.31
MDL Number MFCD00082687
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2514003

  SDS
alfa aesar™
B2514003
1g Each for $71.43

AAB2514006

  SDS
alfa aesar™
B2514006
5g Each for $270.92
Description & Specifications

Specifications

Formula Weight 238.31
Chemical Name or Material 4,5-Diphenyl-1,2,3-thiadiazole
Assay 97%
Molecular Formula C14H10N2S
MDL Number MFCD00082687
CAS 5393-99-7
Melting Point 92° to 94°C
Quantity 1g