Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dimethylthiazole, 98%

Chemical Identifiers

Chemical Name or Material 4,5-Dimethylthiazole
CAS 3581-91-7
Molecular Formula C5H7NS
Formula Weight 113.18
MDL Number MFCD00005336
UN Number UN1993
Beilstein 105694
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1078506

  SDS
alfa aesar™
A1078506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1078514

  SDS
alfa aesar™
A1078514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1078522

  SDS
alfa aesar™
A1078522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.18
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number XJ4380000
Boiling Point 157° to 158°C
Beilstein 105694
Health Hazard 2 GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Molecular Formula C5H7NS
CAS 3581-91-7
Chemical Name or Material 4,5-Dimethylthiazole
Flash Point 51°C (123°F)
UN Number UN1993
Quantity 5g
Assay 98%
Health Hazard 1 Danger
MDL Number MFCD00005336
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.