Please login to your online account to display your discounted pricing

4,5-Dimethyl-2-furaldehyde, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB08889EA

 View more versions of this product

Catalog No. BTB08889EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 124.14
Quantity 10g
Assay 97%
Density 1.0100g/mL
CAS Max % 100.0
Chemical Name or Material 4, 5-Dimethyl-2-furaldehyde
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H8O2
MDL Number MFCD00085030
CAS Min % 96.5
CAS 52480-43-0

Specifications

Chemical Name or Material 4, 5-Dimethyl-2-furaldehyde
CAS 52480-43-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8O2
Formula Weight 124.14
MDL Number MFCD00085030