Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, 98%

$40.16 - $40.16

Chemical Identifiers

Chemical Name or Material 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate
CAS 5808-22-0
Molecular Formula C10H6Na2O8S2·2H2O
Formula Weight 400.29 (364.26 Anhydrous)
MDL Number MFCD00150612
Beilstein 3647189
Merck Index 14,2241
Synonym Chromotropic acid disodium salt dihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1729709

 
alfa aesar™
A1729709
10g Each for $40.16
Description & Specifications

Specifications

Formula Weight 400.29 (364.26 Anhydrous)
Chemical Name or Material 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate
TSCA TSCA
Assay 98%
Molecular Formula C10H6Na2O8S2·2H2O
MDL Number MFCD00150612
CAS 5808-22-0
Beilstein 3647189
Merck Index 14,2241
Synonym Chromotropic acid disodium salt dihydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >300°C
Quantity 10g

Applications
It is employed as a pharmaceutical intermediate and in the preparation of fine chemicals and medicines.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.