Please login to your online account to display your discounted pricing

Alfa Aesar™ 4',5-Dihydroxyflavone, 95%

$89.80 - $804.61

Chemical Identifiers

Chemical Name or Material 4',5-Dihydroxyflavone
CAS 6665-67-4
Molecular Formula C15H10O4
Formula Weight 254.24
MDL Number MFCD00017691
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2765003

  SDS
alfa aesar™
H2765003
1g Each for $89.80

AAH2765006

  SDS
alfa aesar™
H2765006
5g Each for $266.70

AAH2765014

  SDS
alfa aesar™
H2765014
25g Each for $804.61
Description & Specifications

Specifications

Formula Weight 254.24
Molecular Formula C15H10O4
Quantity 1g
Assay 95%
MDL Number MFCD00017691
CAS 6665-67-4
Chemical Name or Material 4',5-Dihydroxyflavone