missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Dihydroxy-4-cyclopentene-1,2,3-trione, 97%, ACROS Organics™

Chemical Identifiers

CAS 488-86-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H2O5
Formula Weight 142.07
MDL Number MFCD00181389
Synonym Croconic acid
Chemical Name or Material 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC374931000 SDS Thermo Fisher Scientific
AC374931000
Glass bottle 100mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

488-86-8
100.0
C5H2O5
MFCD00181389
4,5-Dihydroxy-4-cyclopentene-1,2,3-trione
96.0
97%
142.07
Croconic acid
≥96.0%

Specifications

142.07
100mg
96.0
Croconic acid
MFCD00181389
Glass bottle
488-86-8
≥96.0%
Powder or Crystals
Yellow
100.0
C5H2O5
Authentic
97%
4,5-Dihydroxy-4-cyclopentene-1,2,3-trione

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning