Please login to your online account to display your discounted pricing

4,5-Difluoroanthranilic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398450010

 View more versions of this product

Catalog No. AC398450010

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear colorless to straw
Assay ≥96.0% (HPLC)
Formula Weight 173.12
Physical Form Liquid
Melting Point 180° to 185°C
Quantity 1g
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material 4, 5-Difluoroanthranilic acid
Molecular Formula C7H5F2NO2
Assay Percent Range 97%
CAS 83506-93-8
Linear Formula F2C6H2(NH2)COOH
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00077509
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 4, 5-Difluoroanthranilic acid
CAS 83506-93-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C7H5F2NO2
Linear Formula F2C6H2(NH2)COOH
Formula Weight 173.12
MDL Number MFCD00077509
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents