Please login to your online account to display your discounted pricing

4,5-Difluoroanthranilic acid, 97%, ACROS Organics™

$63.05 - $63.05

Chemical Identifiers

Chemical Name or Material 4, 5-Difluoroanthranilic acid
CAS 83506-93-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C7H5F2NO2
Linear Formula F2C6H2(NH2)COOH
Formula Weight 173.12
MDL Number MFCD00077509
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398450010

  SDS
thermo fisher scientific
AC398450010
Glass bottle 1g Each for $63.05

AC398450050

  SDS
thermo fisher scientific
AC398450050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0% (HPLC)
Formula Weight 173.12
Color Clear colorless to straw
Melting Point 180° to 185°C
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4, 5-Difluoroanthranilic acid
Assay Percent Range 97%
Molecular Formula C7H5F2NO2
Linear Formula F2C6H2(NH2)COOH
CAS 83506-93-8
MDL Number MFCD00077509
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.