missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Difluoro-2-nitrobenzoic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442670010

 View more versions of this product

Catalog No. AC442670010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4, 5-Difluoro-2-nitrobenzoic acid
166.0°C to 167.0°C
20372-63-8
C7H3F2NO4
HGGRAOYTQNFGGN-UHFFFAOYSA-N
203.1
203.1
Glass Bottle
1g
98.5% min. (HPLC)
2-nitro-4,5-difluorobenzoic acid, 3,4-difluoro-6-nitrobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 4,5-two fluoro-2-nitrobenzoic acid, acmc-1cegm, benzoic acid, 4,5-difluoro-2-nitro, pubchem4947
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O
4219630
99%

Chemical Identifiers

20372-63-8
203.1
2-nitro-4,5-difluorobenzoic acid, 3,4-difluoro-6-nitrobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 4,5-two fluoro-2-nitrobenzoic acid, acmc-1cegm, benzoic acid, 4,5-difluoro-2-nitro, pubchem4947
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O
C7H3F2NO4
HGGRAOYTQNFGGN-UHFFFAOYSA-N
4219630
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents