Please login to your online account to display your discounted pricing

4,5-Difluoro-2-nitrobenzoic acid, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4, 5-Difluoro-2-nitrobenzoic acid
Assay Percent Range 99%
Molecular Formula C7H3F2NO4
Formula Weight 203.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC442670050

 
thermo fisher scientific
AC442670050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442670010

 
thermo fisher scientific
AC442670010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.1
Quantity 5g
Chemical Name or Material 4, 5-Difluoro-2-nitrobenzoic acid
Assay Percent Range 99%
Molecular Formula C7H3F2NO4
CAS 20372-63-8