missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,5-Difluoro-2-nitrobenzoic acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442670050
Specifications
4, 5-Difluoro-2-nitrobenzoic acid | |
5g | |
98.5% min. (HPLC) | |
2-nitro-4,5-difluorobenzoic acid, 3,4-difluoro-6-nitrobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 4,5-two fluoro-2-nitrobenzoic acid, acmc-1cegm, benzoic acid, 4,5-difluoro-2-nitro, pubchem4947 | |
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O | |
4219630 | |
99% |
166.0°C to 167.0°C | |
20372-63-8 | |
C7H3F2NO4 | |
HGGRAOYTQNFGGN-UHFFFAOYSA-N | |
203.1 | |
203.1 |
Chemical Identifiers
20372-63-8 | |
203.1 | |
2-nitro-4,5-difluorobenzoic acid, 3,4-difluoro-6-nitrobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 4,5-two fluoro-2-nitrobenzoic acid, acmc-1cegm, benzoic acid, 4,5-difluoro-2-nitro, pubchem4947 | |
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O |
C7H3F2NO4 | |
HGGRAOYTQNFGGN-UHFFFAOYSA-N | |
4219630 |
Safety and Handling
GHS Signal Word: Warning