Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Difluoro-2-nitrobenzoic acid, 97%

$50.92 - $152.19

Chemical Identifiers

Chemical Name or Material 4,5-Difluoro-2-nitrobenzoic acid
CAS 20372-63-8
Molecular Formula C7H3F2NO4
Formula Weight 203.1
MDL Number MFCD00799521
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2893503

  SDS
alfa aesar™
H2893503
1g Each for $50.92

AAH2893506

  SDS
alfa aesar™
H2893506
5g Each for $152.19
Description & Specifications

Specifications

Formula Weight 203.1
Chemical Name or Material 4,5-Difluoro-2-nitrobenzoic acid
Assay 97%
Molecular Formula C7H3F2NO4
MDL Number MFCD00799521
CAS 20372-63-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 163° to 167°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.