missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Difluoro-2-nitroaniline, 98%

Manufacturer:  Alfa Aesar™ H5583006

 View more versions of this product

Catalog No. AAH5583006



Specifications

Specifications

4,5-Difluoro-2-nitroaniline
5g
C6H4F2N2O2
2723242
WDMCABATCGQAKK-UHFFFAOYSA-N
4,5-difluoro-2-nitroaniline
157151
98%
107°C to 108°C
78056-39-0
MFCD00010876
2-amino-4,5-difluoronitrobenzene
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])N
174.107
174.11

Chemical Identifiers

78056-39-0
174.107
WDMCABATCGQAKK-UHFFFAOYSA-N
157151
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])N
C6H4F2N2O2
MFCD00010876
2-amino-4,5-difluoronitrobenzene
4,5-difluoro-2-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents