Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Difluoro-2-methylbenzoic acid, 98%

$140.88 - $352.58

Chemical Identifiers

Chemical Name or Material 4,5-Difluoro-2-methylbenzoic acid
CAS 183237-86-7
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD08062377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2614103

  SDS
alfa aesar™
H2614103
1g Each for $352.58

AAH26141MD

  SDS
alfa aesar™
H26141MD
250mg Each for $140.88
Description & Specifications

Specifications

Formula Weight 172.13
Chemical Name or Material 4,5-Difluoro-2-methylbenzoic acid
Assay 98%
Molecular Formula C8H6F2O2
MDL Number MFCD08062377
CAS 183237-86-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 119° to 123°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.