Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Difluoro-2-methoxybenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 4,5-Difluoro-2-methoxybenzyl bromide
CAS 886499-64-5
Molecular Formula C8H7BrF2O
Formula Weight 237.04
MDL Number MFCD06660220
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2676403

  SDS
alfa aesar™
H2676403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2676406

  SDS
alfa aesar™
H2676406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.04
CAS 886499-64-5
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Molecular Formula C8H7BrF2O
Health Hazard 3 GHS P Statement
P260-P280-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Melting Point 40° to 43°C
MDL Number MFCD06660220
Assay 97%
Health Hazard 1 Danger
Chemical Name or Material 4,5-Difluoro-2-methoxybenzyl bromide
Quantity 1g
UN Number UN3261
Safety and Handling

Danger

GHS P Statement
P260-P280-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.