Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Difluoro-2-methoxybenzonitrile, 99%

Chemical Identifiers

Chemical Name or Material 4,5-Difluoro-2-methoxybenzonitrile
CAS 425702-28-9
Molecular Formula C8H5F2NO
Formula Weight 169.13
MDL Number MFCD06660346
UN Number UN3439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2615603

 
alfa aesar™
H2615603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH26156MD

 
alfa aesar™
H26156MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.13
Chemical Name or Material 4,5-Difluoro-2-methoxybenzonitrile
Assay 99%
Molecular Formula C8H5F2NO
MDL Number MFCD06660346
CAS 425702-28-9
UN Number UN3439
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 98° to 100°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.