Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Difluoro-2-methoxyaniline, 97%

Chemical Identifiers

Chemical Name or Material 4,5-Difluoro-2-methoxyaniline
CAS 1017779-71-3
Molecular Formula C7H7F2NO
Formula Weight 159.14
MDL Number MFCD09832387
UN Number UN2810
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3157503

  SDS
alfa aesar™
H3157503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3157506

  SDS
alfa aesar™
H3157506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.14
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD09832387
UN Number UN2810
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C7H7F2NO
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
CAS 1017779-71-3
Assay 97%
Chemical Name or Material 4,5-Difluoro-2-methoxyaniline
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.