Learn More
4,5-Dicyanoimidazole, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1221506
Description
4,5-Dicyanoimidazole is used in activation of nucleoside phosphoramidites during solid-phase oligonucleotide synthesis, and synthesis of nucleoside phosphoramidites, synthesis of novel nucleosides. It is often used as an alternative to tetrazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5-Dicyanoimidazole is used in activation of nucleoside phosphoramidites during solid-phase oligonucleotide synthesis, and synthesis of nucleoside phosphoramidites, synthesis of novel nucleosides. It is often used as an alternative to tetrazole.
Solubility
Soluble in Acetonitrile: 1.8g/10ml, DMSO, Methanol.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
4,5-Dicyanoimidazole | |
173°C to 177°C | |
C5H2N4 | |
5 g | |
4,5-dicyanoimidazole, imidazole-4,5-dicarbonitrile, 4,5-imidazoledicarbonitrile, activator, pubchem7602, 4, 5-dicyanoimidazole, moa 7, imidazole-4,5-dinitrile, acmc-1brr7, 4,5-dicyano-1h-imidazole | |
XGDRLCRGKUCBQL-UHFFFAOYSA-N | |
1H-imidazole-4,5-dicarbonitrile | |
70729 | |
≥98% |
1122-28-7 | |
Odorless | |
MFCD00005194 | |
118208 | |
Soluble in Acetonitrile: 1.8g/10ml,DMSO,Methanol. | |
N#CC1=C(N=CN1)C#N | |
118.10 | |
118.1 |
Chemical Identifiers
1122-28-7 | |
118.10 | |
XGDRLCRGKUCBQL-UHFFFAOYSA-N | |
1H-imidazole-4,5-dicarbonitrile |
C5H2N4 | |
MFCD00005194 | |
70729 | |
N#CC1=C(N=CN1)C#N |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 214-344-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only