missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dicyano-2-(4-methylphenyl)imidazole, 97%

Manufacturer:  Alfa Aesar™ H5429806

 View more versions of this product

Catalog No. AAH5429806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4,5-Dicyano-2-(4-methylphenyl)imidazole
63857-91-0
MFCD20265230
BMTQOJUWHGWRPD-UHFFFAOYSA-N
2-(4-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
21798437
97%
5g
C12H8N4
2-4-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 2-p-tolyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-4-methylphenyl imidazole, 4,5-dicyano-2-p-tolyl imidazole
CC1=CC=C(C=C1)C2=NC(=C(N2)C#N)C#N
208.224
208.22

Chemical Identifiers

63857-91-0
208.224
BMTQOJUWHGWRPD-UHFFFAOYSA-N
21798437
CC1=CC=C(C=C1)C2=NC(=C(N2)C#N)C#N
C12H8N4
MFCD20265230
2-4-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 2-p-tolyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-4-methylphenyl imidazole, 4,5-dicyano-2-p-tolyl imidazole
2-(4-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents