Alfa Aesar™ 4,5-Dicyano-2-(2-methylphenyl)imidazole, 97%
Manufacturer:  Alfa Aesar™ H54582MD
 View more versions of this product
Catalog No. AAH54582MD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
4,5-Dicyano-2-(2-methylphenyl)imidazole
63857-90-9
MFCD20265294
NQLCQVAWPMBQAB-UHFFFAOYSA-N
2-(2-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
21798430
97%
250mg
C12H8N4
2-2-methylphenyl-1h-imidazole-4,5-dicarbonitrile
CC1=CC=CC=C1C2=NC(=C(N2)C#N)C#N
208.224
208.22
Chemical Identifiers
63857-90-9
208.224
NQLCQVAWPMBQAB-UHFFFAOYSA-N
21798430
CC1=CC=CC=C1C2=NC(=C(N2)C#N)C#N
C12H8N4
MFCD20265294
2-2-methylphenyl-1h-imidazole-4,5-dicarbonitrile
2-(2-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
Safety and Handling
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
Documents