Learn More
4,5-Dichlorophthalic anhydride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 387120050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4, 5-Dichlorophthalic anhydride | |
97.5 | |
185°C to 187°C | |
Authentic | |
Glass bottle | |
MFCD00075034 | |
4,5-dichlorophthalic anhydride, 5,6-dichloroisobenzofuran-1,3-dione, 4,5-dichlorophthalicanhydride, 1,3-isobenzofurandione, 5,6-dichloro, 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione, 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 5,6-dichlor-2-benzofuran-1,3-dion, pubchem16761, acmc-209rqk, 4,5-dichlorophalic anhydride | |
ClC1=CC2=C(C=C1Cl)C(=O)OC2=O | |
217.00 | |
217.01 | |
Powder |
942-06-3 | |
100.0 | |
Blue to Gray | |
97.5% min. (HPLC) | |
C8H2Cl2O3 | |
5 g | |
ULSOWUBMELTORB-UHFFFAOYSA-N | |
5,6-dichloro-2-benzofuran-1,3-dione | |
70334 | |
98% |
Chemical Identifiers
942-06-3 | |
217.00 | |
ULSOWUBMELTORB-UHFFFAOYSA-N | |
70334 | |
ClC1=CC2=C(C=C1Cl)C(=O)OC2=O |
C8H2Cl2O3 | |
MFCD00075034 | |
4,5-dichlorophthalic anhydride, 5,6-dichloroisobenzofuran-1,3-dione, 4,5-dichlorophthalicanhydride, 1,3-isobenzofurandione, 5,6-dichloro, 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione, 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 5,6-dichlor-2-benzofuran-1,3-dion, pubchem16761, acmc-209rqk, 4,5-dichlorophalic anhydride | |
5,6-dichloro-2-benzofuran-1,3-dione |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 213-386-2
RUO – Research Use Only