Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dichloro-2-methylimidazole, 97%

$60.66 - $957.43

Chemical Identifiers

Chemical Name or Material 4,5-Dichloro-2-methylimidazole
CAS 15965-33-0
Molecular Formula C4H4Cl2N2
Formula Weight 151
MDL Number MFCD00051639
Beilstein 606442
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2066703

 
alfa aesar™
B2066703
1g Each for $60.66

AAB2066706

 
alfa aesar™
B2066706
5g Each for $220.12

AAB2066714

 
alfa aesar™
B2066714
25g Each for $957.43
Description & Specifications

Specifications

Formula Weight 151
Chemical Name or Material 4,5-Dichloro-2-methylimidazole
Assay 97%
Molecular Formula C4H4Cl2N2
MDL Number MFCD00051639
CAS 15965-33-0
Beilstein 606442
Melting Point ≈248°C (dec.)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.